missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 3μm Particle Size
A good first choice for compounds with poor peak symmetry on other columns
Brand: Thermo Scientific™ 71603-102130
Code : 16
Additional Details : Weight : 0.10000kg
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
100 mm | |
150 Å | |
C4 | |
Silica | |
Reversed Phase | |
Reversed Phase |
2.1 mm | |
3 μm | |
L26 | |
Beta | |
6% | |
Yes |