missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 3μm Particle Size

A good first choice for compounds with poor peak symmetry on other columns

Brand:  Thermo Scientific™ 71603-051030

Code : 16

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10097594

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • 150Å pore size for small molecules, peptides and protein digests
  • Superb pH stability

Specifications

Specifications

1mm
50mm
3μm
BetaBasic™ 4
Videos