missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 4 HPLC Columns, 3μm Particle Size
A good first choice for compounds with poor peak symmetry on other columns
£472.26 - £517.14
Specifications
Particle Size | 3 μm |
---|---|
Pore Size | 150 Å |
USP Type | L26 |
Stationary Phase | C4 |
Packing Material | Silica |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10753226
|
Thermo Scientific™
71603-052130 |
2.1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10549175
|
Thermo Scientific™
71603-102130 |
2.1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
3 μm | |
L26 | |
Silica | |
Reversed Phase | |
Reversed Phase |
150 Å | |
C4 | |
6% | |
Yes |