Learn More
Thermo Scientific™ ART™ Non-Filtered Multichannel Pipette Tips
A variety of tip styles and configurations
Brand:  Thermo Scientific™ 3897-11
Code : 03
Additional Details : Weight : 7.16000kg
 View more versions of this product
Product Code. 12318542
  • £19.34 / Pack of 250
In Stock 9
Add to basket
Description
Description
The HydroLogix™ (HLT) design utilizes a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipet tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced.
  • Available in blue or clear color
Specifications
Specifications
Bulk, 250 tips
Extended Length
Nonsterile
Bulk
5000μL
Clear
Videos