Learn More
Thermo Scientific™ ART™ Non-Filtered Low Retention Universal Pipette Tips
Specifically designed for eliminating sample retention within the pipette tip
Brand:  Thermo Scientific™ 3581-05
Code : 03
Additional Details : Weight : 4.08230kg
 View more versions of this product
Product Code. 11569984
  • £48.10 / Pack of 800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
  • Crystal clear tips allow you to easily see transparent samples
  • Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting desired volume of liquid
  • All Pure tips are racked, presterilized and certified RNase-, DNase-, DNA- and pyrogen-free
  • Other Thermo Scientific racked and bulk tips are nonsterile and autoclavable
Specifications
Specifications
Rainin™ LTS™ LiteTouch™
Low Retention
Sterile
SoftFit
Rack
100 to 1000μL
Clear
Rainin™ LTS™ LiteTouch™
Videos