Learn More
Thermo Scientific™ Abgene™ Plate Covers
Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking
Brand:  Thermo Scientific™ AB-0752
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11550304
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates. The covers are made of polystyrene or polypropylene and are either ridged or non-ridged.
Highlights
  • Polystyrene covers can be used with all 96-well plates
  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates
Hazardous: No
Storage Condition: Ambient
Specifications
Specifications
Polystyrene
For all 96-well plates
Stacking ridge
Plate Cover
50
Videos
Documentation
Technical Documents