Learn More
Thermo Scientific™ ABgene 0.5mL and 1.0mL Twist-Lock Tube Racks
For sample collection and storage of precious samples including DNA, blood, cell lines, and tissue
Brand: Thermo Scientific™ AB-1312
Code : 04
Additional Details : Weight : 0.80000kg
Alert:
Immersion of tubes and racks in liquid nitrogen introduces safety and contamination risks. Racks should be stored in vapor-phase liquid nitrogen only.
Description
Twist-Lock Tube Racks
- Patent pending internal thread design “locks in” with the cap in two distinct positions, providing robust 100% sealing down to -196°C
- Twist-Lock™ caps are moulded out of a proprietary material which seals more even tightly at cold temperatures
- Lockable racks comply with SBS guidelines and have reduced heights to maximize available freezer space
- All components cleanroom produced
- Suitable for storage at -20°C, -80°C and in vapor-phase liquid nitrogen
- Caps can be applied and removed from tubes while in the rack; easily handled with Twist-Lock Cap-It or Twist-Lock Tool
- Tube base features alphanumeric grid reference corresponding to tube position in the 96-format rack, facilitating sample tracking and identification
- Encapsulated 2D Matrix™ code on each tube base provides robust sample management and tracking; all codes guaranteed to be unique
- 0.5mL tubes: Tube working volume 450μL when capped (400μL recommended when freezing)
- 1.0mL tubes: Tube working volume 900μL when capped (810μL recommended when freezing)
Specifications
Polypropylene | |
For manipulating caps for Twist-Lock tube |