missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ 96 Well 0.8mL Polypropylene Deepwell Storage Plate

0.8 mL deepwell storage plates for cell culture or sample storage applications.

Brand:  Thermo Scientific™ AB-0765/B

Code : 03

Additional Details : Weight : 4.28000kg

 View more versions of this product

Product Code. 10314083

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • 0.8 mL maximum well volume when sealed with adhesive or heat seal
  • 0.70 mL working volume when sealed with Storage Plate Cap Strip
  • 0.55 mL working volume when sealed with a 96-well Sealing Mat
  • Conical well bottom for ease of sample retrieval

 

Compatible Sealing Film: Adhesive Heat
Hazardous: No
Max Centrifugal Force: 2000 g
Max Vol (Cap Seal): 0.7 mL
Max Vol (Film Seal): 0.8 mL
Max Vol (Mat Seal): 0.55 mL
Storage Condition: Ambient

Specifications

Specifications

Blue
Blue
Polypropylene Storage Plate
Videos
Documentation

Technical Documents