missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.2 mL Strip Tubes

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

Brand:  Thermo Scientific™ AB-0451/Y

Code : 03

Additional Details : Weight : 0.26000kg

 View more versions of this product

Product Code. 10712684

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Highlights

  • Compatible with 0.2 mL thermal cycler blocks
  • 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
Specifications

Specifications

8-Tube strip
Certified free from human DNA, RNases, and DNases
Polypropylene
V-Bottom
Domed
Yellow
10 packs of 12 strips
0.2mL
Videos
Documentation

Technical Documents