missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.2 mL Individual Tubes

0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors

Brand:  Thermo Scientific™ AB-0337/B

Code : 03

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10495743

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.

The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed

Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage

Hazardous:No
>Storage Condition:Ambient

PCR; Sample storage

Certifications

Certified free from human DNA, RNases, and DNases

Specifications

Specifications

RNase, Dnase and human DNA free
0.2 mL
0.2mL thermal cycler blocks for PCR and sample storage applications
Domed
Thin Wall PCR Tube
Blue
1000 Case
Polypropylene
Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers