Learn More
Julabo™ Thermal Bath Fluids
Used for temperature control applications. Julabo™ Thermal Bath Fluids offer safe and reliable operation.
Brand: Julabo™ 8940131
Code : A0
Additional Details : Weight : 5.00000kg
Description
- For closed systems like the Presto or Forte
- Has wide temperature ranges
- Low viscosity
- High stability
- Good heat conductivity
- Minimum odor
- Low corrosion tendency
- Low toxicity
- Long shelf life
Specifications
Light Yellow | |
Thermal Bath Fluid | |
Ideal for temperature control applications and guarantee safe and reliable operation, viscosity, oxi | |
H335 | |
373°C |
30°C to 335°C | |
Thermal Bath | |
Drain tap | |
5 L | |
131mm2/s at 20°C |
Safety and Handling
- thermal H335
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : terphenyl