missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium(III) trifluoroacetate, tech.

Brand:  Alfa Aesar™ 043643.18

Code : NEW

Additional Details : CAS Number : 23586-53-0 Weight : 0.05000kg Transport : UN number : 1707 Chem class : 1 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11395857

  • £269.90 / 50g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Thallium (III) trifluoroacetate
Technical Grade
543.42
UN1707
23586-53-0
Tl(OOCCF3)3
MFCD00000414
Thallic trifluoroacetate

Specifications

Thallium (III) trifluoroacetate
Technical Grade
Tl(OOCCF3)3
MFCD00000414
Thallic trifluoroacetate
213°C (dec.)
23586-53-0
Powder
543.42
UN1707
Hygroscopic
50g
Videos
SDS