missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium(III) oxide, 99.9% (metals basis)

In organic synthesis and as an analytical reagent

Brand:  Alfa Aesar™ 043797.09

Code : NEW

Additional Details : CAS Number : 1314-32-5 Weight : 0.01000kg Transport : UN number : 1707 Chem class : 1 Pack group : II

Quantity 10g

 View more versions of this product

Product Code. 11350438

  • £72.80 / 10g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Thallium (III) oxide
Tl2O3
MFCD00011276
14,9268
1314-32-5
456.74
UN1707
Thallium sesquioxide; Thallic oxide

Specifications

Thallium (III) oxide
Powder
(metals basis)
456.74
UN1707
Thallium sesquioxide; Thallic oxide
9.65g/mL
875°C
10g
1314-32-5
99.9%
Tl2O3
MFCD00011276
14,9268
Insoluble in water and alkalies; Decomposes in HCl and H2SO4
Odorless
717°C
Videos
SDS