missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium(I) sulfate, Puratronic™, 99.9995% (metals basis)

Brand:  Alfa Aesar™ 040215.06

Code : NEW

Additional Details : CAS Number : 7446-18-6 Weight : 0.00500kg Transport : UN number : 1707 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11339367

  • £36.15 / 5g


Estimated Shipment date
from Supplier 23-03-2018
Add to basket

Specifications

Chemical Identifiers

Thallium(I) sulfate
Puratronic™
MFCD00011278
14,9269
7446-18-6
504.8
UN1707
Thallous sulfate

Specifications

Thallium(I) sulfate
Puratronic™
99.9995%
504.8
UN1707
Thallous sulfate
Odorless
5g
7446-18-6
Crystalline
(metals basis)
MFCD00011278
14,9269
6.77g/mL
632°C
Videos
SDS