missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) selenide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 11323378
Click to view available options
Quantity:
2 g
This item is not returnable. View return policy
15572-25-5
MFCD00049603
This item is not returnable. View return policy
15572-25-5
MFCD00049603

Thallium(I) selenide is used in molten form as a semiconductor. Semiconducting thallium(I) selenide is used for IR detection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼340°C
Physical Form Lump
Quantity 2 g
UN Number UN1707
Merck Index 14,9267
Solubility Information Insoluble in water and acids.
Formula Weight 487.7
Percent Purity 99.9%
Density 9.05 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) selenide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'