missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) nitrate, 99.5%, Thermo Scientific Chemicals

Catalog Number 10521174
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10102-45-1
NO3Tl
266.38
MFCD00011280
FYWSTUCDSVYLPV-UHFFFAOYSA-N
thallium i nitrate, thallium 1+ nitrate, thallous nitrate, thallium mononitrate, thallium nitrate van, thallium nitrate, tlno3, unii-ljq38dsr12, rcra waste number u217, thallium i nitrate 1:1
24937
thallium(1+);nitrate
[Tl+].[O-][N+]([O-])=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10102-45-1
NO3Tl
266.38
MFCD00011280
FYWSTUCDSVYLPV-UHFFFAOYSA-N
thallium i nitrate, thallium 1+ nitrate, thallous nitrate, thallium mononitrate, thallium nitrate van, thallium nitrate, tlno3, unii-ljq38dsr12, rcra waste number u217, thallium i nitrate 1:1
24937
thallium(1+);nitrate
[Tl+].[O-][N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 433.0°C
Melting Point 206.0°C
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 99.5%
Linear Formula TlNO3
Fieser 13,86
Merck Index 15, 9414
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 266.38
Percent Purity 99.5%
Packaging Glass bottle
Chemical Name or Material Thallium(I) nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'