missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium(I) hexafluoro-2,4-pentanedionate, 99%

Brand:  Alfa Aesar™ 014710.03

Code : NEW

Additional Details : CAS Number : 15444-43-6 Weight : 0.00100kg Transport : UN number : 1707 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11337028

  • £39.01 / 1g


Estimated Shipment date
from Supplier 15-02-2018
Add to basket

Specifications

Chemical Identifiers

Thallium(I) hexafluoro-2,4-pentanedionate
411.42
UN1707
15444-43-6
MFCD00064761
Thallium(I) hexafluoroacetylacetonate

Specifications

Thallium(I) hexafluoro-2,4-pentanedionate
Powder
411.42
UN1707
Yellow
1g
15444-43-6
99%
MFCD00064761
Thallium(I) hexafluoroacetylacetonate
130° to 132°C
Videos
SDS