missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) ethoxide, 95%, Thermo Scientific Chemicals

Catalog Number p-7080349
Click to view available options
Quantity:
5 g
25 g
20398-06-5
C2H5OTl
249.441
MFCD00009072
DZFYOYRNBGNPJW-UHFFFAOYSA-N
thallous ethoxide, thallium i ethanolate, ethoxythallium i, ethanol, thallium 1+ salt, ethyl alcohol, thallium i, ethanol, thallium 1+ salt 1:1, thallium ethoxide, ethanolate; thallium 1+, thallium i ethoxide 5g
30143
ethanolate;thallium(1+)
CC[O-].[Tl+]
20398-06-5
C2H5OTl
249.441
MFCD00009072
DZFYOYRNBGNPJW-UHFFFAOYSA-N
thallous ethoxide, thallium i ethanolate, ethoxythallium i, ethanol, thallium 1+ salt, ethyl alcohol, thallium i, ethanol, thallium 1+ salt 1:1, thallium ethoxide, ethanolate; thallium 1+, thallium i ethoxide 5g
30143
ethanolate;thallium(1+)
CC[O-].[Tl+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 130°C (decomposition)
Melting Point -3°C
Physical Form Liquid
Quantity 5 g
Linear Formula TlOCH2CH3
UN Number UN2810
Flash Point >110°C (230°F)
Formula Weight 249.43
Percent Purity 98%
Refractive Index 1.6714
Sensitivity Moisture sensitive
Density 3.493 g/mL
Chemical Name or Material Thallium (I) ethoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'