missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number 11318577
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7791-12-0
ClTl
239.83
MFCD00011274
GBECUEIQVRDUKB-UHFFFAOYSA-M
thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216
24642
CHEBI:37117
chlorothallium
Cl[Tl]
This item is not returnable. View return policy
7791-12-0
ClTl
239.83
MFCD00011274
GBECUEIQVRDUKB-UHFFFAOYSA-M
thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216
24642
CHEBI:37117
chlorothallium
Cl[Tl]

It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 430°C
Physical Form Beads
Quantity 5 g
UN Number UN1707
Merck Index 14,9260
Solubility Information Slightly soluble in water. Insoluble in alcohol,and acetone.
Formula Weight 239.82
Percent Purity 99.999%
Packaging Ampule
Refractive Index 2.247
Sensitivity Air Sensitive
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'