missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium(I) 2-ethylhexanoate, 99.8% (metals basis)

Brand:  Alfa Aesar™ 036659.04

Code : NEW

Additional Details : CAS Number : 210578-56-6 Weight : 0.00200kg Transport : UN number : 1707 Chem class : 1 Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11348088

  • £14.52 / 2g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Thallium(I) 2-ethylhexanoate
347.59
UN1707
210578-56-6
MFCD00070466

Specifications

Thallium(I) 2-ethylhexanoate
Solid
(metals basis)
MFCD00070466
2g
210578-56-6
99.8%
347.59
UN1707
Videos
SDS