missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Thallium 203, 203Tl, plasma standard solution, Specpure™, 203Tl 10μg/mL

Brand:  Alfa Aesar™ 045288.AD

Code : NEW

Additional Details : Weight : 0.00050kg Transport : UN number : 3264 Chem class : 1a Pack group : III

Quantity 50mL

Product Code. 15443965

  • £91.40 / 50mL


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Specifications

Thallium 203, 203Tl, plasma standard solution
Specpure™
MFCD00084592
50mL
203Tl 10μg/mL
Liquid
UN3264
Matrix: 2% HNO3
Videos
SDS