Learn More
Tocris Bioscience™ TFB-TBOA
High affinity EAAT1 and EAAT2 blocker
Brand:  Tocris Bioscience™ 253210
Code : Z1
Additional Details : Weight : 0.00150kg
 View more versions of this product
Product Code. 13431414
  • £292.53 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Potent and selective glial glutamate transporter EAAT1 and EAAT2 inhibitor (IC50 values are 17, 22 and 300 nM for EAAT2, EAAT1 and EAAT3 respectively). Has no effect on EAAT4 and EAAT5, or a wide range of neuronal receptors and transporters. Attenuates glutamate-stimulated intracellular Na+ elevation in astrocytes in vitro (IC50 = 43 nM). Induces severe convulsions in vivo.
Specifications
Specifications
(3S)-3-[[3-[[4-(Trifluoromethyl)benzoyl]amino]phenyl]methoxy]-L-aspartic acid
>98%
Observed MW: 426.35
Soluble to 100mM in DMSO
480439-73-4
C19H17F3N2O6