Learn More
TFAP4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551658
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: YFMVPTQKVP SLQHFRKTEK EVIGGLCSLA NIPLTPETQR DQERRIRREI ANSNERRRMQ SINAGFQSLK TLIPHTDGEK LSKAAILQQT AEYIFSLEQE KTRLLQQNTQ LKRFIQELSG SSPKRRRAED KDEGIGSPDI WEDEK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
TFAP4 Polyclonal antibody specifically detects TFAP4 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
TFAP4 | |
Polyclonal | |
Unconjugated | |
Tfap4 | |
Activating enhancer-binding protein 4; AI642933; AP4; AP-4; bHLHc41; Class C basic helix-loop-helix protein 41; D930048N17Rik; LOW QUALITY PROTEIN: transcription factor AP-4; Tcfap4; TFAP4; transcription factor AP4; Transcription factor AP-4; transcription factor AP-4 (activating enhancer binding protein 4); transcription factor AP-4 (activating enhancer-binding protein 4) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
360482, 7023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q01664 | |
Tfap4 | |
Recombinant protein corresponding to Human TFAP4 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Research Use Only