Learn More
TFAP2D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561847
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TPAICAALST FQTVLSEMLN YLEKHTTHKN GGAADSGQGH ANSEKAPLRK TSEAAVKEGK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
TFAP2D Polyclonal antibody specifically detects TFAP2D in Human samples. It is validated for Immunohistochemistry.Specifications
TFAP2D | |
Polyclonal | |
Unconjugated | |
TFAP2D | |
activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; activating enhancer-binding protein 2-delta; AP-2 like; AP-2-delta; AP2-delta; hypothetical protein LOC566838; si:dkey-5e16.2; Tcfap2d; TFAP2BL1; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 beta-like 1; transcription factor AP-2 delta; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); transcription factor AP-2, delta; transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta; zgc:114196 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83741 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6R9 | |
TFAP2D | |
Recombinant protein corresponding to Human TFAP2D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only