Learn More
Texwipe™ TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
For ISO Class 6 and 7 applications
Brand:  Texwipe™ TX604
Additional Details : Weight : 4.53590kg
 View more versions of this product
Product Code. 11595335
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Economical all-purpose nonwoven wipers made from a blend of 55% cellulose and 45% polyester—combine the absorbency of natural fiber with the cleanliness and strength of synthetic fiber.
  • Hydroentanglement process uses jets of water to blend fibers, eliminating need for chemical binders
  • Provide excellent bidirectional strength and very low solvent extractables levels
  • Offer superior wet and dry strength, solvent-compatibility and acid resistance
  • Will not scratch or abrade delicate precision components
Specifications
Specifications
360
69
4 x 4 in. (10.2 x 10.16 cm)
Designed for general purpose wiping where economy and cleanliness are of critical importance, ideal for applications where high absorbency and low extractables are prime considerations
Cellulose, Polyester
Yes
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
10.16 cm
10.2 cm
TechniCloth™ dry wipes
Videos