Learn More
Texwipe™ TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
For ISO Class 6 and 7 applications
Brand: Texwipe™ TX606
Additional Details : Weight : 5.66990kg
Description
Economical all-purpose nonwoven wipers made from a blend of 55% cellulose and 45% polyester—combine the absorbency of natural fiber with the cleanliness and strength of synthetic fiber.
- Hydroentanglement process uses jets of water to blend fibers, eliminating need for chemical binders
- Provide excellent bidirectional strength and very low solvent extractables levels
- Offer superior wet and dry strength, solvent-compatibility and acid resistance
- Will not scratch or abrade delicate precision components
Specifications
360 | |
69 | |
6 x 6 in. (15.24 x 15.24 cm) | |
Designed for general purpose wiping where economy and cleanliness are of critical importance, ideal for applications where high absorbency and low extractables are prime considerations | |
Cellulose, Polyester |
Yes | |
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999 | |
15.24 cm | |
15.24 cm | |
TechniCloth™ dry wipes |