Learn More
Texwipe™ Sterile TechniSat™ Presaturated Wipers
For ISO Class 6 applications
Brand:  Texwipe™ TX3217
Code : 04
Additional Details : Weight : 8.20000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
 View more versions of this product
Product Code. 12662560
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Binder-free hydroentangled polyester/cellulose wiper presaturated with USP-grade 70% isopropyl alcohol 30% deionized water, filtered through 0.2μm filter
  • Gamma irradiated to 10-6 Sterility Assurance Level in accordance with Association for Advancement of Medical Instrumentation
  • Low levels of solvent extractables and particle generation
Specifications
Specifications
11 x 9 in. (28 x 23 cm)
70% Isopropyl Alcohol, 30% Deionised Water
23 cm
28 cm
Presaturated Wipers
20 wipers/Flexpack
Yes
Designed for cleaning and polishing equipment and environmental surfaces during and following the production flow
Polyester, Cellulose
Videos