Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Apply directly to floor
Brand:  Texwipe™ AMA254581G
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11515325
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
Blue
36 x 24 in.
60 layer
Videos