Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 60 Layer
Apply directly to floor
Brand:  Texwipe™ AMA364682G
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11515335
  • £425.00 / Pack of 4
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Choose 30- or 60-layer mats.
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
Gray
46 x 36 in.
60 layer
Videos