Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Brand:  Texwipe™ AMA364681G
Additional Details : Weight : 16.78290kg
 View more versions of this product
Product Code. 11545325
  • £486.00 / Pack of 8
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
Gray
46 x 36 in.
30 layer
Videos