Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Brand:  Texwipe™ AMA364681W
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11535325
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
White
46 x 36 in.
30 layer
Videos