Learn More
Texwipe™ AlphaMop™
Flat mopping system for easy and efficient cleaning of floors, walls and ceilings
Brand:  Texwipe™ TX7118
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11551224
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Specifically designed for the needs of ultraclean production environments. Easily changeable, laundered polyester mop covers fit over a flat, foam-padded head assembly. Lightweight, durable fiberglass handle provides excellent control over swivel head and accesses hard-to-reach areas with ease.
  • Enables rapid change of wiping surfaces for adherence to strict wiping protocol ISO Class 4 cleanroom-laundered Alpha™ polyester mop cover
  • Replaceable foam pad ensures that the polyester mop cover conforms to textured floors and other surfaces
  • Maintains ultralow generation of particles and extractables Resists corrosion, abrasion and commonly used cleaning and disinfecting chemicals such as bleaches, phenols and quaternary ammonium compound
  • AlphaMop polyester replacement mopheads and foam pads available separately (150 heads and six pads per case)
  • AlphaMop mop covers presaturated with 6% IPA Honeycomb™ 10 double-knit polyester mop covers have an abrasive texture (supplied 150 covers per case)
Specifications
Specifications
Polyester
No
50 mop covers and 6 pads
Replacement Mop Head and Foam Pad
Designed for the needs of ultraclean production environments
Videos