Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
Brand:  Texwipe™ TX7114
Code : 04
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12356337
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
  • Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
  • Made from autoclavable Celcon™ plastic
  • Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Specifications
For cleanroom class 4 to 8
Size: 4 x 7 in.
Yes
Mini AlphaMop™ AlphaWipe™ replacement mop covers
Replacement mop covers only
150/Cs.
Polyester
Videos