Learn More
Texwipe™ TexTra™ Wipers
Thick, cleanroom polyester knit wipers for rigorous wiping
Brand: Texwipe™ TX3423
Code : NEW
Additional Details : Weight : 0.01000kg
Beschreibung
- These single-ply wipers have the liquid-holding capacity of two-ply wipers
- TexTra™ combines this high absorbency with great abrasion resistance
- Can be autoclaved for sterile or aseptic environments
Spezifikation
ISO Class 3 – 8, Class 1 – 100,000, EU Grade A – D | |
25 x 60 cm | |
Sterile |