missing translation for 'onlineSavingsMsg'
Learn More

Tetrindole mesylate, Tocris Bioscience™

MAO-A inhibitor

Brand:  Tocris (Bio-Techne) 072310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13487893

  • £127.00 / 10mg


Estimated Shipment
03-07-2018
Add to basket

Description

Description

Novel antidepressant, a selective inhibitor of MAO-A.
Specifications

Chemical Identifiers

Tetrindole mesylate
C20H26N2·CH3SO3H
390.54
2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate

Specifications

Tetrindole mesylate
170964-68-8
C20H26N2·CH3SO3H
2,3,3a,4,5,6-Hexahydro-8-cyclohexyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
10mg
>99%
390.54
Videos