missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate, 95%

Brand:  Alfa Aesar™ 030538.03

Code : NEW

Additional Details : CAS Number : 122451-08-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11317308

  • £251.40 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate
1239.11 (1022.87 Anhydrous)
5488862
122451-08-5
MFCD00151011
meso-Tetra(sulfonatophenyl)porphine tetrasodium salt dodecahydrate

Specifications

Tetrasodium meso-tetra(sulfonatophenyl)porphine dodecahydrate
122451-08-5
95%
MFCD00151011
meso-Tetra(sulfonatophenyl)porphine tetrasodium salt dodecahydrate
1g
Crystalline
1239.11 (1022.87 Anhydrous)
5488862
Videos
SDS