missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium perruthenate, ACROS Organics™

Brand:  Acros Organics 378390010

Code : 29

Additional Details : CAS Number : 114615-82-6 Weight : 0.05100kg Transport : UN number : 1479 Chem class : 1 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10662065

  • £71.90 / 1g


EU Stock 11
Add to basket


Specifications

Chemical Identifiers

Tetrapropylammonium perruthenate
C12H28NO4Ru
MFCD00074914
114615-82-6
351.43

Specifications

Tetrapropylammonium perruthenate
160.0°C
114615-82-6
Glass bottle
C12H28NO4Ru
MFCD00074914
15, 9386
Black
1g
Authentic
Powder
351.43
27.7% min.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrapropylammonium perruthenate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS