missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium perchlorate, 98%, ACROS Organics™

Brand:  Acros Organics 420741000

Code : 29

Additional Details : CAS Number : 15780-02-6 Weight : 0.15000kg Transport : UN number : 1479 Chem class : 1 Pack group : II


Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10484895

  • £243.30 / 100g


Estimated Shipment
09-02-2018
Add to basket


Specifications

Chemical Identifiers

Tetrapropylammonium perchlorate
98%
(CH3CH2CH2)4NClO4
MFCD00041979
≥97.5% (on dry substance)
C12H28ClNO4
285.8

Specifications

Tetrapropylammonium perchlorate
100g
Authentic
Chunks
98%
(CH3CH2CH2)4NClO4
MFCD00041979
White
15780-02-6
Plastic bottle
≥97.5% (on dry substance)
C12H28ClNO4
285.8
2 to 10%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrapropylammonium perchlorate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS