missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropylammonium perchlorate, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagent, Phase transfer catalyst
Specifications
Melting Point | 238°C to 240°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (on dry substance) |
Packaging | Plastic bottle |
Linear Formula | (CH3CH2CH2)4NClO4 |
Quantity | 25 g |
Formula Weight | 285.8 |
Percent Purity | 98% |
Physical Form | Chunks |
Show More |
Safety and Handling
Product Identifier
- Tetrapropylammonium perchlorate
- Danger
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'