missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylphthalic anhydride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7081834
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
4741-53-1
C32H20O3
452.509
MFCD00044504
RSKXGCFFFZIWNC-UHFFFAOYSA-N
tetraphenylphthalic anhydride, 4,5,6,7-tetraphenylisobenzofuran-1,3-dione, acmc-20ajiw, tetraphenylphthalicanhydride, 3,4,5,6-tetraphenylphthalic anhydride, tetraphenyl-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, 4,5,6,7-tetraphenyl
78478
4,5,6,7-tetraphenyl-2-benzofuran-1,3-dione
C1=CC=C(C=C1)C2=C(C(=C3C(=C2C4=CC=CC=C4)C(=O)OC3=O)C5=CC=CC=C5)C6=CC=CC=C6
This item is not returnable. View return policy
4741-53-1
C32H20O3
452.509
MFCD00044504
RSKXGCFFFZIWNC-UHFFFAOYSA-N
tetraphenylphthalic anhydride, 4,5,6,7-tetraphenylisobenzofuran-1,3-dione, acmc-20ajiw, tetraphenylphthalicanhydride, 3,4,5,6-tetraphenylphthalic anhydride, tetraphenyl-2-benzofuran-1,3-dione, 1,3-isobenzofurandione, 4,5,6,7-tetraphenyl
78478
4,5,6,7-tetraphenyl-2-benzofuran-1,3-dione
C1=CC=C(C=C1)C2=C(C(=C3C(=C2C4=CC=CC=C4)C(=O)OC3=O)C5=CC=CC=C5)C6=CC=CC=C6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 289°C to 297°C
Quantity 5 g
Beilstein 353504
Sensitivity Moisture sensitive
Formula Weight 452.51
Percent Purity 98%
Chemical Name or Material Tetraphenylphthalic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'