missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylphosphonium bromide, 98+%, Thermo Scientific Chemicals
22.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
100 g
Description
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Incompatible with strong oxidizing agents. Stable at room temperature in closed containers under normal storage and handling conditions.
Tetraphenylphosphonium bromide is a supporting electrolyte for the electroreduction of buckminsterfullerene. It is used to extract heavy metals from aqueous solution as ion-association complexes
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Incompatible with strong oxidizing agents. Stable at room temperature in closed containers under normal storage and handling conditions.
Specifications
Melting Point | 296°C to 302°C |
Quantity | 10 g |
Beilstein | 3922383 |
Sensitivity | Hygroscopic |
Merck Index | 14,9237 |
Solubility Information | Soluble in water. |
Formula Weight | 419.31 |
Percent Purity | ≥98% |
Chemical Name or Material | Tetraphenylphosphonium bromide |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'