missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tetraoctadecylammonium bromide, 98%

Brand:  Alfa Aesar™ 020582.03

Code : NEW

Additional Details : CAS Number : 63462-99-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11317957

  • £80.56 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Tetraoctadecylammonium bromide
63462-99-7
1107.9
MFCD00043172

Specifications

Tetraoctadecylammonium bromide
103° to 104°C
63462-99-7
98%
MFCD00043172
Hygroscopic
1g
Crystalline
1107.9
Videos
SDS