missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyltin, 98%, ACROS Organics™

£49.04 - £213.93

Chemical Identifiers

CAS 594-27-4
Molecular Formula C4H12Sn
Molecular Weight (g/mol) 178.83
MDL Number MFCD00008278
InChI Key VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Synonym tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
PubChem CID 11661
ChEBI CHEBI:30420
IUPAC Name tetramethylstannane
SMILES C[Sn](C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10001950 SDS View Documents Acros Organics
163980100
10g Glass bottle £49.04
10g View alternative products
Save Up to 
»
Estimated Shipment
19-08-2020
Please to purchase this item. Need a web account? Register with us today!
10265590 SDS View Documents Acros Organics
163980500
50g Glass bottle £213.93
50g View alternative products
Save Up to 
»
Estimated Shipment
04-08-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

594-27-4
178.83
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
11661
tetramethylstannane
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
CHEBI:30420
C[Sn](C)(C)C

Specifications

Tetramethyltin
−12°C
Glass bottle
1.29
Colorless
10g
594-27-4
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
tetramethylstannane
11661
178.83
98%
1.2900g/mL
Authentic
1.4400 to 1.4420
74.0°C to 75.0°C
-54.0°C
99%
97.5% min. (GC)
(CH3)4Sn
04, 631
Solubility in water: immiscible.
C[Sn](C)(C)C
178.83
CHEBI:30420
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • Tetramethyltin

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.