missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyltetraselenafulvalene, 97%, Thermo Scientific Chemicals

Catalog Number 10712885
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
55259-49-9
C10H12Se4
448.04
MFCD00011507
YMWLPMGFZYFLRP-UHFFFAOYSA-N
tetramethyltetraselenafulvalene, tmtsf, 1,3-diselenole, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl-1,3-diselenole, tetramethyltetraselenafulvalene, purum, 4,4',5,5'-tetramethyl-.delta.2,2'-bi-1,3-diselenole, 1,3-diselenole, 4,5-dimethyl-2-4,5-dimethyl-1,3-diselenol-2-ylidene
143265
2-(4,5-dimethyl-1,3-diselenol-2-ylidene)-4,5-dimethyl-1,3-diselenole
CC1=C([Se]C(=C2[Se]C(=C([Se]2)C)C)[Se]1)C
This item is not returnable. View return policy
55259-49-9
C10H12Se4
448.04
MFCD00011507
YMWLPMGFZYFLRP-UHFFFAOYSA-N
tetramethyltetraselenafulvalene, tmtsf, 1,3-diselenole, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl, 2-4,5-dimethyl-1,3-diselenol-2-ylidene-4,5-dimethyl-1,3-diselenole, tetramethyltetraselenafulvalene, purum, 4,4',5,5'-tetramethyl-.delta.2,2'-bi-1,3-diselenole, 1,3-diselenole, 4,5-dimethyl-2-4,5-dimethyl-1,3-diselenol-2-ylidene
143265
2-(4,5-dimethyl-1,3-diselenol-2-ylidene)-4,5-dimethyl-1,3-diselenole
CC1=C([Se]C(=C2[Se]C(=C([Se]2)C)C)[Se]1)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 265.0°C to 270.0°C
Color Red-Violet
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 448.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Tetramethyltetraselenafulvalene, 97%
compliance-icons compliance-icons
Product Identifier
  • Tetramethyltetraselenafulvalene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'