Learn More
Tetramethyltetraselenafulvalene, 97%, ACROS Organics™
Brand: Acros Organics 346500500
Code : 29
Additional Details : CAS Number : 55259-49-9 Weight : 0.05005kg Transport : UN number : 3283 Chem class : 1 Pack group : II
Quantity | 50mg |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
Tetramethyltetraselenafulvalene | |
55259-49-9 | |
C10H12Se4 | |
MFCD00011507 |
97% | |
97% | |
448.04 |
Specifications
Tetramethyltetraselenafulvalene | |
265.0° to 270.0°C | |
97% | |
Complies, 96.0% min. | |
Crystalline Powder | |
97% | |
448.04 |
Red-violet | |
50mg | |
55259-49-9 | |
Glass bottle | |
97% | |
C10H12Se4 | |
MFCD00011507 |