missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals

Catalog Number p-4627721
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
75-76-3
C4H12Si
88.23
MFCD00008274
CZDYPVPMEAXLPK-UHFFFAOYSA-N
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
6396
CHEBI:85361
tetramethylsilane
C[Si](C)(C)C
This item is not returnable. View return policy
75-76-3
C4H12Si
88.23
MFCD00008274
CZDYPVPMEAXLPK-UHFFFAOYSA-N
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
6396
CHEBI:85361
tetramethylsilane
C[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -99°C
Color Colorless
Density 0.6400g/mL
Boiling Point 26°C to 28°C
Flash Point -27°C
Packaging Glass bottle
Linear Formula Si(CH3)4
Refractive Index 1.3585
Quantity 25 g
Merck Index 15,9377
Specific Gravity 0.64
Solubility Information Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 88.22
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Tetramethylsilane
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'