missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylcyclopentadiene, mixed isomers, tech. 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tetramethylcyclopentadiene is used in pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.808 g/mL |
Boiling Point | 55°C to 59°C (13 mmHg) |
Flash Point | 42°C (108°F) |
Refractive Index | 1.472 |
Quantity | 1 g |
UN Number | UN3295 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Not miscible mix with mater. |
Formula Weight | 122.21 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'