missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 98%, Acros Organics

£22.40 - £338.00

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H13NO·5H2O
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES C[N+](C)(C)C.O.O.O.O.O.[OH-]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10474895 SDS View Documents Acros Organics
420530050
5g Glass bottle £22.40
5g View alternative products
Save Up to 
»
EU Stock 11 Please to purchase this item. Need a web account? Register with us today!
10647761 SDS View Documents Acros Organics
420530250
25g Glass Bottle £29.00
25g View alternative products
Save Up to 
»
EU Stock 14 Please to purchase this item. Need a web account? Register with us today!
10657761 SDS View Documents Acros Organics
420531000
100g Glass Bottle £78.90
100g View alternative products
Save Up to 
»
In Stock 5
Please to purchase this item. Need a web account? Register with us today!
10388193 SDS View Documents Acros Organics
420535000
500g Glass bottle £338.00
500g View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

10424-65-4
181.23
MYXKPFMQWULLOH-UHFFFAOYSA-M
82620
C[N+](C)(C)C.O.O.O.O.O.[OH-]
C4H13NO·5H2O
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
tetramethylazanium;hydroxide;pentahydrate

Specifications

Tetramethylammonium hydroxide pentahydrate
Glass bottle
62.0°C to 71.0°C
10424-65-4
C4H13NO·5H2O
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium;hydroxide;pentahydrate
82620
Crystalline Powder
Authentic
White
5g
98%
(CH3)4NOH·5H2O
04, 50
Solubility in water: soluble.
C[N+](C)(C)C.O.O.O.O.O.[OH-]
181.23
181.23
98%
Safety and Handling

Safety and Handling

Product Identifier
  • Tetramethylammonium hydroxide pentahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.