missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 97%, Thermo Scientific Chemicals

Catalog Number p-3737472
24.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10424-65-4
C4H23NO6
181.23
MFCD00149566
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
82620
tetramethylazanium;hydroxide;pentahydrate
O.O.O.O.O.[OH-].C[N+](C)(C)C
This item is not returnable. View return policy
10424-65-4
C4H23NO6
181.23
MFCD00149566
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
82620
tetramethylazanium;hydroxide;pentahydrate
O.O.O.O.O.[OH-].C[N+](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Melting Point 62.0°C to 71.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CH3)4NOH·5H2O
Quantity 25 g
Beilstein 04, 50
Solubility Information Solubility in water: soluble.
Formula Weight 181.23
Percent Purity 97%
Physical Form Hygroscopic Crystals
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'