Learn More
Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 420510010
81.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 29
Additional Details : CAS Number : 75-59-2 Weight : 1.00000kg Transport : UN number : 1835 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetramethylammonium hydroxide | |
75-59-2,7732-18-5 | |
1.0000g/mL | |
Plastic bottle | |
(CH3)4NOH | |
1 kg | |
11,514 | |
15, 9371 | |
Solubility in water: soluble. | |
C[N+](C)(C)C.[OH-] | |
91.15 | |
91.15 | |
Liquid |
10% in Water | |
Colorless | |
100.0°C | |
C4H13NO | |
MFCD00008280 | |
04, 50 | |
1 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
9.5 to 10.5% |
RUO – Research Use Only